missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxy-2-nitrobenzoic Acid 97.0+%, TCI America™
$63.38 - $249.78
Chemical Identifiers
| CAS | 1882-69-5 |
|---|---|
| Molecular Formula | C8H6NO5 |
| Molecular Weight (g/mol) | 196.14 |
| MDL Number | MFCD00151836 |
| InChI Key | URADKXVAIGMTEG-UHFFFAOYSA-M |
| Synonym | 5-methoxy-2-nitrobenzoic acid, 2-nitro-5-methoxybenzoic acid, 6-nitro-m-anisic acid, 5-methoxy-2-nitro-benzoic acid, benzoic acid, 5-methoxy-2-nitro, pubchem4229, acmc-1brd6, 2-nitro-5-methoxybenzoate, 2-nitro-5-methoxybenzoicacid, 5-methoxy-2-nitrobenzoicacid |
| PubChem CID | 339209 |
| IUPAC Name | 5-methoxy-2-nitrobenzoate |
| SMILES | COC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O |
Chemical Identifiers
| 1882-69-5 | |
| 196.14 | |
| URADKXVAIGMTEG-UHFFFAOYSA-M | |
| 339209 | |
| COC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O |
| C8H6NO5 | |
| MFCD00151836 | |
| 5-methoxy-2-nitrobenzoic acid, 2-nitro-5-methoxybenzoic acid, 6-nitro-m-anisic acid, 5-methoxy-2-nitro-benzoic acid, benzoic acid, 5-methoxy-2-nitro, pubchem4229, acmc-1brd6, 2-nitro-5-methoxybenzoate, 2-nitro-5-methoxybenzoicacid, 5-methoxy-2-nitrobenzoicacid | |
| 5-methoxy-2-nitrobenzoate |
Specifications
| 1882-69-5 | |
| Yellow | |
| MFCD00151836 | |
| 5-methoxy-2-nitrobenzoic acid, 2-nitro-5-methoxybenzoic acid, 6-nitro-m-anisic acid, 5-methoxy-2-nitro-benzoic acid, benzoic acid, 5-methoxy-2-nitro, pubchem4229, acmc-1brd6, 2-nitro-5-methoxybenzoate, 2-nitro-5-methoxybenzoicacid, 5-methoxy-2-nitrobenzoicacid | |
| COC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O | |
| 196.14 | |
| 197.15 | |
| Crystalline Powder |
| 132°C | |
| C8H6NO5 | |
| 5 g | |
| URADKXVAIGMTEG-UHFFFAOYSA-M | |
| 5-methoxy-2-nitrobenzoate | |
| 339209 | |
| ≥97.0% (GC,T) | |
| 5-Methoxy-2-nitrobenzoic Acid |
Safety and Handling
TSCA : No