missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-nitroaniline, 98%

Catalog No. AAH3404603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3404603 1 g
AAH3404609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3404603 Supplier Thermo Scientific Chemicals Supplier No. H3404603

CAS: 16133-49-6 | C7H8N2O3 | 168.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16133-49-6
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00179573
InChI Key QEHVRGACCVLLNN-UHFFFAOYSA-N
Synonym 5-methoxy-2-nitrophenylamine, 3-amino-4-nitroanisole, 5methoxy-2-nitroaniline, 5-methoxy-2-nitrobenzenamine, 3-amino-4-nitro-anisol, 2-nitro-5-methoxyaniline, 5-methoxy-2-nitro-aniline, benzenamine, 5-methoxy-2-nitro, benzenamine,5-methoxy-2-nitro, 5-methoxy-2-nitro-phenylamine
PubChem CID 85300
IUPAC Name 5-methoxy-2-nitroaniline
SMILES COC1=CC(=C(C=C1)[N+](=O)[O-])N

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 5-Methoxy-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 240-293-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only