missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxy-2-methylindole 99.0+%, TCI America™
$227.16 - $899.05
Chemical Identifiers
| CAS | 1076-74-0 |
|---|---|
| Molecular Formula | C10H11NO |
| Molecular Weight (g/mol) | 161.20 |
| MDL Number | MFCD00005620 |
| InChI Key | VSWGLJOQFUMFOQ-UHFFFAOYSA-N |
| Synonym | 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole |
| PubChem CID | 70642 |
| IUPAC Name | 5-methoxy-2-methyl-1H-indole |
| SMILES | COC1=CC=C2NC(C)=CC2=C1 |
Chemical Identifiers
| 1076-74-0 | |
| 161.20 | |
| VSWGLJOQFUMFOQ-UHFFFAOYSA-N | |
| 70642 | |
| COC1=CC=C2NC(C)=CC2=C1 |
| C10H11NO | |
| MFCD00005620 | |
| 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole | |
| 5-methoxy-2-methyl-1H-indole |
Specifications
| 1076-74-0 | |
| White-Yellow | |
| C10H11NO | |
| 1 g | |
| VSWGLJOQFUMFOQ-UHFFFAOYSA-N | |
| 5-methoxy-2-methyl-1H-indole | |
| 70642 | |
| ≥99.0% (GC) | |
| 5-Methoxy-2-methylindole |
| 87°C | |
| 145°C | |
| MFCD00005620 | |
| 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole | |
| COC1=CC=C2NC(C)=CC2=C1 | |
| 161.20 | |
| 161.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No