missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylindole 99.0+%, TCI America™

Catalog No. M07305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07305G 5 g
M07301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07305G Supplier TCI America Supplier No. M07305G

Chemical Identifiers

CAS 1076-74-0
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005620
InChI Key VSWGLJOQFUMFOQ-UHFFFAOYSA-N
Synonym 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole
PubChem CID 70642
IUPAC Name 5-methoxy-2-methyl-1H-indole
SMILES COC1=CC=C2NC(C)=CC2=C1

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 161.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxy-2-methylindole