missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylindole, 99+%

Catalog No. AAB2134803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2134803 1 g
AAB2134806 5 g
AAB2134814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2134803 Supplier Thermo Scientific Chemicals Supplier No. B2134803

CAS: 1076-74-0 | C10H11NO | 161.20 g/mol

It is employed as a reactant in preparation of indolylquinoxalines by condensation reactions, reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation, reactant in arylation reactions using a palladium acetate catalyst, reactant in enantioselective Friedel-Crafts alkylation and reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation. As an indole derivative shown to be an effective inhibitor of the chlorinating activity of myeloperoxidase (MPO). Used in the metabolic synthesis of arylacetic acid anti-inflammatory synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant in preparation of indolylquinoxalines by condensation reactions, reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation, reactant in arylation reactions using a palladium acetate catalyst, reactant in enantioselective Friedel-Crafts alkylation and reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation. As an indole derivative shown to be an effective inhibitor of the chlorinating activity of myeloperoxidase (MPO). Used in the metabolic synthesis of arylacetic acid anti-inflammatory synthesis.

Solubility
Soluble in methanol (very faint turbidity.)

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1076-74-0
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005620
InChI Key VSWGLJOQFUMFOQ-UHFFFAOYSA-N
Synonym 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole
PubChem CID 70642
IUPAC Name 5-methoxy-2-methyl-1H-indole
SMILES COC1=CC=C2NC(C)=CC2=C1

Specifications

Melting Point 85°C to 88°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in methanol (very faint turbidity.)
Formula Weight 161.2
Percent Purity ≥99%
Chemical Name or Material 5-Methoxy-2-methylindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-066-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only