missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 50868-72-9 |
|---|---|
| Molecular Formula | C8H11NO |
| Molecular Weight (g/mol) | 137.18 |
| MDL Number | MFCD00075057 |
| InChI Key | RPJXLEZOFUNGNZ-UHFFFAOYSA-N |
| Synonym | 5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic |
| PubChem CID | 99500 |
| IUPAC Name | 5-methoxy-2-methylaniline |
| SMILES | COC1=CC=C(C)C(N)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC398240050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 50868-72-9 | |
| 137.18 | |
| RPJXLEZOFUNGNZ-UHFFFAOYSA-N | |
| 99500 | |
| COC1=CC=C(C)C(N)=C1 |
| C8H11NO | |
| MFCD00075057 | |
| 5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic | |
| 5-methoxy-2-methylaniline |
Specifications
| 50868-72-9 | |
| 100.0 | |
| Authentic | |
| C8H11NO | |
| MFCD00075057 | |
| RPJXLEZOFUNGNZ-UHFFFAOYSA-N | |
| 5-methoxy-2-methylaniline | |
| 99500 | |
| 97% |
| 96.0 | |
| 113°C | |
| 97% | |
| CH3OC6H3(CH3)NH2 | |
| 5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic | |
| COC1=CC=C(C)C(N)=C1 | |
| 137.18 | |
| 137.18 | |
| 5-Methoxy-2-methylaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 256-816-4
RUO – Research Use Only