missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1-tetralone 98.0+%, TCI America™

Catalog No. p-7133142
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22975G 5 g
M229725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22975G Supplier TCI America Supplier No. M22975G

Chemical Identifiers

CAS 33892-75-0
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001692
InChI Key BRCPWISABURVIH-UHFFFAOYSA-N
Synonym 5-methoxy-1-tetralone, 5-methoxy-3,4-dihydronaphthalenone, 5-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-5-methoxy, 5-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, alpha-tetralone, 5-methoxy, 3,4-dihydro-5-methoxy-1 2h-naphthalenone, 5-methoxy-2,3,4-trihydronaphthalen-1-one, 5-methoxytetralone, pubchem17899
PubChem CID 36620
IUPAC Name 5-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC=CC2=C1CCCC2=O

Specifications

Melting Point 90°C
Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxy-1-tetralone
RTECSNumber QK4800000
TSCA No