missing translation for 'onlineSavingsMsg'
Learn More

5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt, Thermo Scientific™

Catalog No. p-7059223
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4421904 2 g
AA4421909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4421904 Supplier Thermo Scientific Chemicals Supplier No. 4421904
Only null left

As a complexing reagent for Bi, Te, Se and other metal ions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6336-51-2
Molecular Formula C8H5KN2S3
Molecular Weight (g/mol) 264.42
MDL Number MFCD00014092
InChI Key QSKDCVGFAPHSHX-UHFFFAOYSA-M
Synonym 1,3,4-thiadiazolidine-2,5-dithione, 3-phenyl-, potassium salt, 3-phenyl-1,3,4-thiadiazolidine-2,5-dithione potassium, potassium 5-mercapto-3-phenyl-1,3,4-thiadiazole-2-thione, 3-phenyl-5-sulfanyl-1,3,4-thiadiazole-2-thione potassium, 5-mercapto-3-phenyl-1,3,4-thiadiazole-2 3h thione potassium salt
PubChem CID 2775614
IUPAC Name potassium;4-phenyl-5-sulfanylidene-1,3,4-thiadiazole-2-thiolate
SMILES C1=CC=C(C=C1)N2C(=S)SC(=N2)[S-].[K+]

Specifications

Quantity 2 g
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
Formula Weight 264.43
Physical Form Powder
Chemical Name or Material 5-Mercapto-3-phenyl-1,3,4-thiadiazole-2 (3H)thione potassium salt
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-718-1
TSCA No
Recommended Storage Ambient temperatures