missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Mercapto-1H-1,2,3-triazole Sodium Salt 97.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 123.11 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Mercapto-1H-1,2,3-triazole Sodium Salt |
Safety and Handling
| EINECSNumber | (5)-5827 |
| TSCA | No |