accessibility menu, dialog, popup

UserName

CAS RN 59032-27-8

IUPAC Name: sodium (2H-1,2,3-triazol-4-yl)sulfanide dihydrate
Synonyms: sodium 2H-1,2,3-triazol-4-ylsulfanide dihydrate
Molecular Weight (g/mol): 159.14
Molecular Formula: C2H6N3NaO2S
InChi Key: RZLGEIPBTHKAGX-UHFFFAOYSA-M
SMILES: O.O.[Na+].[S-]C1=NNN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results