missing translation for 'onlineSavingsMsg'
Learn More

5-Mercapto-1H-1,2,3-triazole Sodium Salt 97.0+%, TCI America™

Catalog No. M08475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08475G 5 g
M084725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08475G Supplier TCI America Supplier No. M08475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59032-27-8
Molecular Formula C2H2N3NaS
Molecular Weight (g/mol) 123.109
MDL Number MFCD00044799
InChI Key HHGMADGROXARPN-UHFFFAOYSA-M
Synonym sodium 1,2,3-triazole-5-thiolate, 5-mercapto-1,2,3-triazole monosodium salt, sodium 1h-1,2,3-triazol-5-ylsulfanide, 1h-1,2,3-triazole-5-thiol sodium salt, 1h-5-mercapto-1,2,3-triazole,sodium salt, tas:1h-1,2,3-triazole-4-thiol sodium salt, sodium 1h-1,2,3-triazole-4-thiolate, sodium 1h-1,2,3-triazole-5-thiolate, sodium 2h-1,2,3-triazole-4-thiolate, 1h-1,2,3-triazole-4-thiol sodium salt
PubChem CID 23685427
IUPAC Name sodium;2H-triazole-4-thiolate
SMILES C1=NNN=C1[S-].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 123.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Mercapto-1H-1,2,3-triazole Sodium Salt
EINECSNumber (5)-5827
TSCA No