missing translation for 'onlineSavingsMsg'
Learn More

5-Mercapto-1-methyltetrazole, 98%

Catalog No. AAB2044306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2044306 5 g
AAB2044314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2044306 Supplier Thermo Scientific Chemicals Supplier No. B2044306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13183-79-4 | C2H4N4S | 116.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13183-79-4
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00037317
InChI Key XOHZHMUQBFJTNH-UHFFFAOYSA-N
Synonym 5-mercapto-1-methyltetrazole, 1-methyl-5-mercaptotetrazole, 1-methyl-1h-tetrazole-5-thiol, 1-n-methyl-5-thiotetrazole, 1-methyltetrazole-5-thiol, n-methyltetrazolethiol, 1-methyl-5-tetrazolethione, 1-methyl-5-mercapto-1h-tetrazole, 1-methyl-5-mercapto-1,2,3,4-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-methyl
PubChem CID 2723772
IUPAC Name 1-methyl-2H-tetrazole-5-thione
SMILES CN1NN=NC1=S

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Formula Weight 116.14
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 5-Mercapto-1-methyltetrazole
Hazard Category Warning
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement GHS P Statement

P261-P280-P305+P351+P338-P304+P340-P405-P501a

Avoid breathing dust/fume/gas/mist/vapors/spray.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Store locked up.

Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 236-132-2
RTECSNumber XF7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only