accessibility menu, dialog, popup

UserName

CH3NNHNNS

CAS RN 13183-79-4

IUPAC Name: 1-methyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms: 1-methyl-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol): 116.14
Molecular Formula: C2H4N4S
InChi Key: XOHZHMUQBFJTNH-UHFFFAOYSA-N
SMILES: CN1NN=NC1=S
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)