missing translation for 'onlineSavingsMsg'
Learn More

5-(m-Tolyl)-5H-pyrrolo[3,2-c:4,5-c']dipyridine 95.0+%, TCI America™

Catalog No. t3281100mg
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
T3281100MG 100 mg
1 options

Catalog No. T3281100MG

Supplier: TCI America T3281100MG

Only null left

Chemical Identifiers

CAS 1014403-09-8
Molecular Formula C17H13N3
Molecular Weight (g/mol) 259.312
InChI Key WAQDNMLJAYQJRN-UHFFFAOYSA-N
PubChem CID 59546683
SMILES CC1=CC(=CC=C1)N2C3=C(C=NC=C3)C4=C2C=CN=C4

Specifications

Quantity 100 mg
Formula Weight 259.31
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(m-Tolyl)-5H-pyrrolo[3,2-c:4,5-c′]dipyridine