missing translation for 'onlineSavingsMsg'
Learn More

5'-m-Terphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T27921G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27921G Supplier TCI America Supplier No. T27921G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128388-54-5
Molecular Formula C18H15BO2
Molecular Weight (g/mol) 274.126
MDL Number MFCD09953491
InChI Key MRBZYVMZUBUDAX-UHFFFAOYSA-N
Synonym 3,5-diphenylphenyl boronic acid, 1,1':3',1-terphenyl-5'-ylboronic acid, 1,1':3',1-terphenyl-5'-boronic acid, 3,5-diphenylbenzeneboronic acid, 5'-m-terphenylboronic acid, boronic acid, 1,1':3',1-terphenyl-5'-yl, boronic acid,b-1,1':3',1-terphenyl-5'-yl, 5-phenyl-1,1'-biphenyl-3-ylboronic acid, 3,5-diphenylphenyl, acmc-1cc4x
PubChem CID 14739363
IUPAC Name (3,5-diphenylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3)(O)O

Specifications

Melting Point 296°C
Color White
Quantity 1 g
Formula Weight 274.13
Physical Form Crystalline Powder
Chemical Name or Material 5′-m-Terphenylboronic Acid (contains varying amounts of Anhydride)