missing translation for 'onlineSavingsMsg'
Learn More

5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™

$199.95 - $199.95

Chemical Identifiers

CAS 551930-53-1
Molecular Formula C7H4ClNO3S2
Molecular Weight (g/mol) 249.68
InChI Key SSGKBJJYLTYNQD-UHFFFAOYSA-N
Synonym 5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
PubChem CID 2795330
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC16003DA
SDS View Documents
Thermo Scientific Chemicals
CC16003DA
1 g
Each for $199.95
Only null left
 
Description

Description

Specifications

Chemical Identifiers

551930-53-1
249.68
5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
C7H4ClNO3S2
SSGKBJJYLTYNQD-UHFFFAOYSA-N
2795330

Specifications

551930-53-1
1 g
SSGKBJJYLTYNQD-UHFFFAOYSA-N
2795330
Tech.
5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
C7H4ClNO3S2
5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
249.68
249.7
Technical
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'