CAS RN 551930-53-1
5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™
Chemical Name or Material: 5-Isoxazol-5-ylthiophene-2-sulfonyl chloride Molecular Formula: C7H4ClNO3S2 Formula Weight: 249.7
5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™
CAS: 551930-53-1 Molecular Formula: C7H4ClNO3S2 Molecular Weight (g/mol): 249.68 InChI Key: SSGKBJJYLTYNQD-UHFFFAOYSA-N Synonym: 5-5-isoxazyl thiophene-2-sulfonyl chloride,5-isoxazol-5-ylthiophene-2-sulfonyl chloride,5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride,5-5-isoxazolyl-2-thiophenesulfonyl chloride,5-isoxazol-5-yl thiophene-2-sulfonyl chloride,chloro 5-isoxazol-5-yl 2-thienyl sulfone,ssgkbjjyltynqd-uhfffaoysa-n,5-5-isoxazyl thiophene-2-sulphonyl chloride,5-5-isoxazolyl-2-thiophenesulfonoyl chloride,5-5-isoxazolyl thiophene-2-sulfonyl chloride PubChem CID: 2795330