missing translation for 'onlineSavingsMsg'
Learn More

5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™

Catalog No. cc16003da
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC16003DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC16003DA Supplier Thermo Scientific Chemicals Supplier No. CC16003DA
Only null left

Chemical Identifiers

CAS 551930-53-1
Molecular Formula C7H4ClNO3S2
Molecular Weight (g/mol) 249.68
InChI Key SSGKBJJYLTYNQD-UHFFFAOYSA-N
Synonym 5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
PubChem CID 2795330

Specifications

Quantity 1 g
Formula Weight 249.7
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
Hazard Category GHS Signal Word: Danger