missing translation for 'onlineSavingsMsg'
Learn More

5-Isopropyl-m-xylene 85.0+%, TCI America™

Catalog No. I0156500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0156500ML 500 mL
I015625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0156500ML Supplier TCI America Supplier No. I0156500ML

Chemical Identifiers

CAS 4706-90-5
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00026405
InChI Key RMKJTYPFCFNTGQ-UHFFFAOYSA-N
Synonym 3,5-Dimethylcumene, 3,5-Dimethylisopropylbenzene
PubChem CID 20833
IUPAC Name 1,3-dimethyl-5-(propan-2-yl)benzene
SMILES CC(C)C1=CC(C)=CC(C)=C1

Specifications

Color Colorless
Boiling Point 195°C
Quantity 500 mL
UN Number 3295
Formula Weight 148.25
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Isopropyl-m-xylene
EINECSNumber (3)-3427
TSCA No