missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodovanillin 98.0+%, TCI America™
$59.65 - $160.30
Chemical Identifiers
| CAS | 5438-36-8 |
|---|---|
| Molecular Formula | C8H7IO3 |
| Molecular Weight (g/mol) | 278.045 |
| MDL Number | MFCD00006941 |
| InChI Key | FBBCSYADXYILEH-UHFFFAOYSA-N |
| Synonym | 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68 |
| PubChem CID | 79499 |
| IUPAC Name | 4-hydroxy-3-iodo-5-methoxybenzaldehyde |
| SMILES | COC1=C(C(=CC(=C1)C=O)I)O |
Chemical Identifiers
| 5438-36-8 | |
| 278.045 | |
| FBBCSYADXYILEH-UHFFFAOYSA-N | |
| 79499 | |
| COC1=C(C(=CC(=C1)C=O)I)O |
| C8H7IO3 | |
| MFCD00006941 | |
| 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68 | |
| 4-hydroxy-3-iodo-5-methoxybenzaldehyde |
Specifications
| 5438-36-8 | |
| Yellow | |
| MFCD00006941 | |
| 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68 | |
| COC1=C(C(=CC(=C1)C=O)I)O | |
| 278.045 | |
| 278.05 | |
| Crystalline Powder |
| 182°C | |
| C8H7IO3 | |
| 5 g | |
| FBBCSYADXYILEH-UHFFFAOYSA-N | |
| 4-hydroxy-3-iodo-5-methoxybenzaldehyde | |
| 79499 | |
| ≥98.0% (GC) | |
| 5-Iodovanillin |
Safety and Handling
TSCA : Yes