missing translation for 'onlineSavingsMsg'
Learn More

5-Iodovanillin 98.0+%, TCI America™

Catalog No. I04275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04275G 5 g
I042725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04275G Supplier TCI America Supplier No. I04275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5438-36-8
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00006941
InChI Key FBBCSYADXYILEH-UHFFFAOYSA-N
Synonym 5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
PubChem CID 79499
IUPAC Name 4-hydroxy-3-iodo-5-methoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)I)O

Specifications

Melting Point 182°C
Color Yellow
Quantity 5 g
Formula Weight 278.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodovanillin