missing translation for 'onlineSavingsMsg'
Learn More

5-Iodouridine, 96%

Catalog No. AAJ6225903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6225903 1 g
AAJ6225906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6225903 Supplier Thermo Scientific Chemicals Supplier No. J6225903

CAS: 1024-99-3 | C9H11IN2O6 | 370.099 g/mol

5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 1024-99-3
Molecular Formula C9H11IN2O6
Molecular Weight (g/mol) 370.099
MDL Number MFCD00006532
InChI Key RKSLVDIXBGWPIS-UAKXSSHOSA-N
Synonym 5-iodouridine, uridine, 5-iodo, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidine-2,4 1h,3h-dione, 5-iodo-uridine, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-iodo-3h-pyrimidine-2,4-dione, 5-i-uridine, 2,4-dihydroxy-5-iodo-1-beta-d-ribofuranosylpyrimidine, ksc179a6b, uridine, 5-iodo-8ci 9ci
PubChem CID 1268108
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)I

Specifications

Color White
Quantity 1 g
Beilstein 33665
Solubility Information Soluble in water.
Formula Weight 370.1
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 5-Iodouridine
EINECSNumber 213-833-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only