missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Iodouracil, TCI America™

$24.44 - $67.22

Chemical Identifiers

CAS 696-07-1
Molecular Formula C4H3IN2O2
Molecular Weight (g/mol) 237.984
MDL Number MFCD00006020
InChI Key KSNXJLQDQOIRIP-UHFFFAOYSA-N
Synonym 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 69672
ChEBI CHEBI:43636
IUPAC Name 5-iodo-1H-pyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I021925G View Documents TCI America
I021925G
25g Each for $67.22
 
I02195G View Documents TCI America
I02195G
5g Each for $24.44
 
Specifications

Chemical Identifiers

696-07-1
237.984
KSNXJLQDQOIRIP-UHFFFAOYSA-N
69672
5-iodo-1H-pyrimidine-2,4-dione
C4H3IN2O2
MFCD00006020
5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
CHEBI:43636
C1=C(C(=O)NC(=O)N1)I

Specifications

White
696-07-1
MFCD00006020
KSNXJLQDQOIRIP-UHFFFAOYSA-N
5-iodo-1H-pyrimidine-2,4-dione
69672
237.98
≥99.0% (HPLC,T)
25g
C4H3IN2O2
5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
C1=C(C(=O)NC(=O)N1)I
237.984
CHEBI:43636
Crystal-Powder at 20°C
5-Iodouracil
Safety and Handling

Safety and Handling

RTECSNumber : YR0525000

TSCA : Yes