missing translation for 'onlineSavingsMsg'
Learn More

5-Iodouracil 99.0+%, TCI America™

Supplier:  TCI America I02195G

 View more versions of this product

Catalog No. I02195G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Iodouracil
White
MFCD00006020
5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
IC1=CNC(=O)NC1=O
237.98
CHEBI:43636
≥99.0% (HPLC,T)
696-07-1
C4H3IN2O2
5 g
KSNXJLQDQOIRIP-UHFFFAOYSA-N
5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
69672
237.98
Crystalline Powder

Chemical Identifiers

696-07-1
237.98
KSNXJLQDQOIRIP-UHFFFAOYSA-N
69672
5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
C4H3IN2O2
MFCD00006020
5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
CHEBI:43636
IC1=CNC(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YR0525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions