missing translation for 'onlineSavingsMsg'
Learn More

5-Iodosalicylic Acid 98.0+%, TCI America™

Catalog No. I053825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I053825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I053825G Supplier TCI America Supplier No. I053825G

Chemical Identifiers

CAS 119-30-2
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.018
MDL Number MFCD00002458
InChI Key SWDNKOFGNPGRPI-UHFFFAOYSA-N
Synonym 5-iodosalicylic acid, benzoic acid, 2-hydroxy-5-iodo, salicylic acid, 5-iodo, 5-iodo-2-hydroxybenzoic acid, 2-hydroxy-5-iodo-benzoic acid, rarechem al be 0824, labotest-bb lt00454718, 5-jod-salicylsaure, 5-iodosalicylicacid, 5-iodo-salicylicaci
PubChem CID 8388
IUPAC Name 2-hydroxy-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)O

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 25 g
Formula Weight 264.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodosalicylic Acid
RTECSNumber VO4200000
TSCA No