missing translation for 'onlineSavingsMsg'
Learn More

5-Iodosalicylic acid, 97%

Catalog No. aab2129136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2129136 500 g
AAB2129114 25 g
AAB2129122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2129136 Supplier Thermo Scientific Chemicals Supplier No. B2129136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 119-30-2 | C7H5IO3 | 264.018 g/mol

2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 119-30-2
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.018
MDL Number MFCD00002458
InChI Key SWDNKOFGNPGRPI-UHFFFAOYSA-N
Synonym 5-iodosalicylic acid, benzoic acid, 2-hydroxy-5-iodo, salicylic acid, 5-iodo, 5-iodo-2-hydroxybenzoic acid, 2-hydroxy-5-iodo-benzoic acid, rarechem al be 0824, labotest-bb lt00454718, 5-jod-salicylsaure, 5-iodosalicylicacid, 5-iodo-salicylicaci
PubChem CID 8388
IUPAC Name 2-hydroxy-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)O

Specifications

Melting Point 194°C to 199°C
Quantity 500 g
Beilstein 2090443
Formula Weight 264.02
Percent Purity 97%
Chemical Name or Material 5-Iodosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-313-5
RTECSNumber VO4200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only