missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodo-m-xylene 98.0+%, TCI America™
$111.84 - $335.52
Chemical Identifiers
| CAS | 22445-41-6 |
|---|---|
| Molecular Formula | C8H9I |
| Molecular Weight (g/mol) | 232.06 |
| MDL Number | MFCD00060659 |
| InChI Key | ZLMKEENUYIUKKC-UHFFFAOYSA-N |
| Synonym | 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, # |
| PubChem CID | 140924 |
| IUPAC Name | 1-iodo-3,5-dimethylbenzene |
| SMILES | CC1=CC(I)=CC(C)=C1 |
Chemical Identifiers
| 22445-41-6 | |
| 232.06 | |
| ZLMKEENUYIUKKC-UHFFFAOYSA-N | |
| 140924 | |
| CC1=CC(I)=CC(C)=C1 |
| C8H9I | |
| MFCD00060659 | |
| 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, # | |
| 1-iodo-3,5-dimethylbenzene |
Specifications
| 22445-41-6 | |
| 229°C | |
| Clear Liquid at 20°C | |
| MFCD00060659 | |
| ZLMKEENUYIUKKC-UHFFFAOYSA-N | |
| 1-iodo-3,5-dimethylbenzene | |
| 140924 | |
| ≥98.0% (GC) |
| Yellow | |
| 5 g | |
| C8H9I | |
| 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, # | |
| CC1=CC(I)=CC(C)=C1 | |
| 232.06 | |
| 232.06 | |
| 5-Iodo-m-xylene |
Safety and Handling
TSCA : No