missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-m-xylene 98.0+%, TCI America™

Catalog No. I05485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05485G 5 g
I054825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05485G Supplier TCI America Supplier No. I05485G

Chemical Identifiers

CAS 22445-41-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00060659
InChI Key ZLMKEENUYIUKKC-UHFFFAOYSA-N
Synonym 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, #
PubChem CID 140924
IUPAC Name 1-iodo-3,5-dimethylbenzene
SMILES CC1=CC(I)=CC(C)=C1

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 229°C
Formula Weight 232.06
Percent Purity ≥98.0% (GC)
Chemical Name or Material 5-Iodo-m-xylene