missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-m-xylene, 97%

Catalog No. AAA1841309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1841309 10 g
AAA1841318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1841309 Supplier Thermo Scientific Chemicals Supplier No. A1841309

CAS: 22445-41-6 | C8H9I | 232.06 g/mol

1-Iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is suitable for use in the synthesis of N-(3,5-xylyl)-N-ethyl aniline, an aryl amine, It is also used in the following studies; α-Arylation of ketones, Copper-catalyzed N-arylation of imidazoles, Cyanation of 5-iodo-m-xylene to form 3,5-dimethylbenzonitrile, synthesis of 1,3-Dimethyl-5-phenoxybenzene by nano-CuFe2O4 catalyzed C-O cross-coupling with phenol, CuBr-catalyzed amination of 1-iodo-3,5-dimethylbenzene to form N-Allyl-3,5-dimethylbenzenamine, copper-catalyzed C-S bond-formation between 5-iodo-m-xylene and thiophenol. It is also employed as a starting material in the synthesis of biphenyl-3,3’,5,5’-tetracarboxylic acid, radical bromination of 5-iodo-m-xylene by N-bromosuccinimide to form 1,3-bis(bromomethyl)-5-iodobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is suitable for use in the synthesis of N-(3,5-xylyl)-N-ethyl aniline, an aryl amine, It is also used in the following studies; α-Arylation of ketones, Copper-catalyzed N-arylation of imidazoles, Cyanation of 5-iodo-m-xylene to form 3,5-dimethylbenzonitrile, synthesis of 1,3-Dimethyl-5-phenoxybenzene by nano-CuFe2O4 catalyzed C-O cross-coupling with phenol, CuBr-catalyzed amination of 1-iodo-3,5-dimethylbenzene to form N-Allyl-3,5-dimethylbenzenamine, copper-catalyzed C-S bond-formation between 5-iodo-m-xylene and thiophenol. It is also employed as a starting material in the synthesis of biphenyl-3,3’,5,5’-tetracarboxylic acid, radical bromination of 5-iodo-m-xylene by N-bromosuccinimide to form 1,3-bis(bromomethyl)-5-iodobenzene.

Solubility
Sparingly soluble (0.014 g/L) (25°C), Calc.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 22445-41-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00060659
InChI Key ZLMKEENUYIUKKC-UHFFFAOYSA-N
Synonym 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, #
PubChem CID 140924
IUPAC Name 1-iodo-3,5-dimethylbenzene
SMILES CC1=CC(I)=CC(C)=C1

Specifications

Density 1.61
Quantity 10 g
Refractive Index 1.595
Beilstein 2039353
Boiling Point 229°C
Solubility Information Sparingly soluble (0.014g/L) (25°C),Calc.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 5-Iodo-m-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only