missing translation for 'onlineSavingsMsg'
Learn More

5'-Iodo-m-terphenyl 98.0+%, TCI America™

Catalog No. I0969200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0969200MG 200 mg
I09691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0969200MG Supplier TCI America Supplier No. I0969200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87666-86-2
Molecular Formula C18H13I
Molecular Weight (g/mol) 356.206
MDL Number MFCD26131291
InChI Key LICOFADCKUKHJI-UHFFFAOYSA-N
Synonym 1-Iodo-3,5-diphenylbenzene, 5′C-Iodo-1,1′C:3′C,1′C′C-terphenyl
PubChem CID 13067230
IUPAC Name 1-iodo-3,5-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2)I)C3=CC=CC=C3

Specifications

Melting Point 103°C
Quantity 200 mg
Formula Weight 356.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Iodo-m-terphenyl