missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1,2,3-trimethoxybenzene, 97%

CAS: 25245-29-8 | C9H11IO3 | 294.09 g/mol

$149.33 - $582.55

Chemical Identifiers

CAS 25245-29-8
Molecular Formula C9H11IO3
Molecular Weight (g/mol) 294.09
MDL Number MFCD01318153
InChI Key IWPMQXOVTMABLF-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyiodobenzene, benzene, 5-iodo-1,2,3-trimethoxy, acmc-1cfcc, 3,4,5-trimethoxy-iodobenzene, 3,4,5-trimethoxyphenyl iodide, 3,4,5-trimethoxy-iodo-benzene, 1-iodo-3,4,5-trimethoxybenzene, 1,2,3-trimethoxy-5-iodobenzene, 5-iodo-1,2,3-trimethoxy-benzene, 5-iodanyl-1,2,3-trimethoxy-benzene
PubChem CID 298133
IUPAC Name 5-iodo-1,2,3-trimethoxybenzene
SMILES COC1=CC(I)=CC(OC)=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1741403
View Documents
Thermo Scientific Chemicals
L1741403
1 g
Each for $149.33
Only null left
 
AAL1741406
View Documents
Thermo Scientific Chemicals
L1741406
5 g
Each for $582.55
Only null left
 
Description

Description

5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

25245-29-8
294.09
IWPMQXOVTMABLF-UHFFFAOYSA-N
298133
COC1=CC(I)=CC(OC)=C1OC
C9H11IO3
MFCD01318153
3,4,5-trimethoxyiodobenzene, benzene, 5-iodo-1,2,3-trimethoxy, acmc-1cfcc, 3,4,5-trimethoxy-iodobenzene, 3,4,5-trimethoxyphenyl iodide, 3,4,5-trimethoxy-iodo-benzene, 1-iodo-3,4,5-trimethoxybenzene, 1,2,3-trimethoxy-5-iodobenzene, 5-iodo-1,2,3-trimethoxy-benzene, 5-iodanyl-1,2,3-trimethoxy-benzene
5-iodo-1,2,3-trimethoxybenzene

Specifications

25245-29-8
C9H11IO3
1 g
Light sensitive
Insoluble in water.
COC1=CC(I)=CC(OC)=C1OC
294.09
294.09
5-Iodo-1,2,3-trimethoxybenzene
85°C to 87°C
MFCD01318153
1529690
3,4,5-trimethoxyiodobenzene, benzene, 5-iodo-1,2,3-trimethoxy, acmc-1cfcc, 3,4,5-trimethoxy-iodobenzene, 3,4,5-trimethoxyphenyl iodide, 3,4,5-trimethoxy-iodo-benzene, 1-iodo-3,4,5-trimethoxybenzene, 1,2,3-trimethoxy-5-iodobenzene, 5-iodo-1,2,3-trimethoxy-benzene, 5-iodanyl-1,2,3-trimethoxy-benzene
IWPMQXOVTMABLF-UHFFFAOYSA-N
5-iodo-1,2,3-trimethoxybenzene
298133
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only