missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxytryptamine, MP Biomedicals™
$323.39 - $323.39
Chemical Identifiers
| CAS | 153-98-0 |
|---|---|
| Molecular Formula | C10H13ClN2O |
| Molecular Weight (g/mol) | 212.677 |
| InChI Key | MDIGAZPGKJFIAH-UHFFFAOYSA-N |
| Synonym | serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride |
| PubChem CID | 160436 |
| IUPAC Name | 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride |
| SMILES | C1=CC2=C(C=C1O)C(=CN2)CCN.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15376550
|
MP Biomedicals Inc
0215376550 |
50 mg |
Each for $323.39
|
|
|||||
Chemical Identifiers
| 153-98-0 | |
| 212.677 | |
| serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride | |
| 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride |
| C10H13ClN2O | |
| MDIGAZPGKJFIAH-UHFFFAOYSA-N | |
| 160436 | |
| C1=CC2=C(C=C1O)C(=CN2)CCN.Cl |
Specifications
| 153-98-0 | |
| 0.993 | |
| 50 mg | |
| MDIGAZPGKJFIAH-UHFFFAOYSA-N | |
| 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride | |
| 160436 | |
| Powder |
| Gray | |
| C10H13ClN2O | |
| serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride | |
| C1=CC2=C(C=C1O)C(=CN2)CCN.Cl | |
| 212.677 | |
| 169.2°C - 170.6°CC | |
| 5-Hydroxytryptamine |