missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyomeprazole, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015908
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015908 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015908 Supplier Merck Emd Millipore Supplier No. 439995MG
Only null left
Pharmaceutical (small molecule)

Specifications

CAS 92340-57-3
Physical Form Neat
Linear Formula C17H19N3O4 S
MDL Number MFCD00869771
Synonym 4-Methoxy-6-{[(5-methoxy-1 H-benzimidazol-2-yl)sulfinyl]methyl}-5-methyl-3-pyridinemethanol; 5-Methoxy-2-{[(5-hydroxymethyl-4-methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl}-1 H-benzimidazole
Type Analytical Standard
Quantity 5 mg
Formula Weight 361.42
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard