missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethyl-2-furaldehyde (stabilized with Water) 95.0+%, TCI America™

Catalog No. H02691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02691G 1 g
H02695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02691G Supplier TCI America Supplier No. H02691G

Chemical Identifiers

CAS 67-47-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003234
InChI Key NOEGNKMFWQHSLB-UHFFFAOYSA-N
Synonym 5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
PubChem CID 237332
ChEBI CHEBI:412516
IUPAC Name 5-(hydroxymethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C=O)CO

Specifications

Color Brown-Yellow
Quantity 1 g
Formula Weight 126.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hydroxymethyl-2-furaldehyde (stabilized with Water)
RTECSNumber LT7031100
TSCA Yes
Recommended Storage Refrigerator