missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxyisoquinoline 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 220°C |
| Quantity | 5 g |
| Formula Weight | 145.16 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Hydroxyisoquinoline |
Safety and Handling
| TSCA | No |