missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole 98.0+%, TCI America™

Catalog No. p-7130444
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0252100MG 100 mg
H02521G 1 g
H02525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0252100MG Supplier TCI America Supplier No. H0252100MG
Only null left

Chemical Identifiers

CAS 1953-54-4
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005677
InChI Key LMIQERWZRIFWNZ-UHFFFAOYSA-N
Synonym 5-hydroxyindole, indol-5-ol, 5-hydroxy-1h-indole, hydroxy-5 indole, 5-hydroxy-indole, 5-indolol, hydroxy-5-indole, 5-hydroxyindol, 5-hydroxy indole, unii-320un7xzyn
PubChem CID 16054
IUPAC Name 1H-indol-5-ol
SMILES OC1=CC=C2NC=CC2=C1

Specifications

Melting Point 107°C
Quantity 100 mg
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyindole
RTECSNumber NM2430000
TSCA No