accessibility menu, dialog, popup

UserName

OHHN

CAS RN 1953-54-4

IUPAC Name: 1H-indol-5-ol
Synonyms: 5-hydroxyindol
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: LMIQERWZRIFWNZ-UHFFFAOYSA-N
SMILES: OC1=CC=C2NC=CC2=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)