missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole, 98+%

Catalog No. AAL0085806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0085806 5 g
AAL0085803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0085806 Supplier Thermo Scientific Chemicals Supplier No. L0085806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1953-54-4 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1953-54-4
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005677
InChI Key LMIQERWZRIFWNZ-UHFFFAOYSA-N
Synonym 5-hydroxyindole, indol-5-ol, 5-hydroxy-1h-indole, hydroxy-5 indole, 5-hydroxy-indole, 5-indolol, hydroxy-5-indole, 5-hydroxyindol, 5-hydroxy indole, unii-320un7xzyn
PubChem CID 16054
IUPAC Name 1H-indol-5-ol
SMILES OC1=CC=C2NC=CC2=C1

Specifications

Melting Point 106°C to 111°C
Quantity 5 g
Beilstein 112349
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material 5-Hydroxyindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-782-6
RTECSNumber NM2430000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only