missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic Acid 98.0+%, TCI America™

Catalog No. H03551G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03551G 1 g
H0355100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03551G Supplier TCI America Supplier No. H03551G

Chemical Identifiers

CAS 54-16-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00005639
InChI Key DUUGKQCEGZLZNO-UHFFFAOYSA-N
Synonym 5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
PubChem CID 1826
ChEBI CHEBI:27823
IUPAC Name 2-(5-hydroxy-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1O)C(=CN2)CC(=O)O

Specifications

Melting Point 162°C
Color Purple-White
Quantity 1 g
Formula Weight 191.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyindole-3-acetic Acid
RTECSNumber NL3650000
TSCA No