missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic acid, 98%

Catalog No. AAJ61435ME
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61435ME 500 mg
AAJ6143503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61435ME Supplier Thermo Scientific Chemicals Supplier No. J61435ME

Metabolite of melatonin | CAS: 54-16-0 | C10H9NO3 | 191.186 g/mol

5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.

Solubility
Soluble in ethanol (50 mg/ml).

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 54-16-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00005639
InChI Key DUUGKQCEGZLZNO-UHFFFAOYSA-N
Synonym 5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
PubChem CID 1826
ChEBI CHEBI:27823
IUPAC Name 2-(5-hydroxy-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1O)C(=CN2)CC(=O)O

Specifications

Melting Point 161°C to 164°C (decomposition)
Color Purple to White
Quantity 500 mg
Beilstein 168797
Sensitivity Light sensitive
Solubility Information Soluble in ethanol (50mg/ml).
Formula Weight 191.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyindole-3-acetic acid
EINECSNumber 200-195-4
RTECSNumber NL3650000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only