missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindan 99.0+%, TCI America™

Catalog No. H025125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H025125G 25 g
H0251100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H025125G Supplier TCI America Supplier No. H025125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1470-94-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003802
InChI Key PEHSSTUGJUBZBI-UHFFFAOYSA-N
Synonym 5-indanol, indan-5-ol, 5-hydroxyindan, 5-hydroxyhydrindene, 5-hydroxyindane, 1h-inden-5-ol, 2,3-dihydro, indanol-5, unii-9z94h6f15t, 5-indano
PubChem CID 15118
ChEBI CHEBI:59311
IUPAC Name 2,3-dihydro-1H-inden-5-ol
SMILES OC1=CC=C2CCCC2=C1

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 134.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyindan
RTECSNumber NK7525000
TSCA Yes