missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyflavone, 97%

CAS: 491-78-1 | C15H10O3 | 238.24 g/mol

Supplier:  Thermo Scientific Chemicals L14187ME

 View more versions of this product

Catalog No. AAL14187ME

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Hydroxyflavone
156°C to 160°C
MFCD00016944
194429
IYBLVRRCNVHZQJ-UHFFFAOYSA-N
5-hydroxy-2-phenylchromen-4-one
68112
97%
491-78-1
C15H10O3
500 mg
5-hydroxyflavone, primuletin, 5-hydroxy-2-phenyl-4h-chromen-4-one, 5-hydroxy-2-phenyl-chromen-4-one, chembl16807, 4h-1-benzopyran-4-one,5-hydroxy-2-phenyl, primuliten, 5-hydroxy-flavone, 5-hydroxy-2-phenyl-4-benzopyrone, 5-hydroxy-2-phenyl-4h-chromen-4-on
OC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1
238.24
238.25

Chemical Identifiers

491-78-1
238.24
IYBLVRRCNVHZQJ-UHFFFAOYSA-N
68112
OC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1
C15H10O3
MFCD00016944
5-hydroxyflavone, primuletin, 5-hydroxy-2-phenyl-4h-chromen-4-one, 5-hydroxy-2-phenyl-chromen-4-one, chembl16807, 4h-1-benzopyran-4-one,5-hydroxy-2-phenyl, primuliten, 5-hydroxy-flavone, 5-hydroxy-2-phenyl-4-benzopyrone, 5-hydroxy-2-phenyl-4h-chromen-4-on
5-hydroxy-2-phenylchromen-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-743-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only