accessibility menu, dialog, popup

UserName

CAS RN 491-78-1

IUPAC Name: 5-hydroxy-2-phenyl-4H-chromen-4-one
Synonyms: 5-hydroxyflavone primuletin
Molecular Weight (g/mol): 238.24
Molecular Formula: C15H10O3
InChi Key: IYBLVRRCNVHZQJ-UHFFFAOYSA-N
SMILES: OC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)