missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxydiclofenac, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 69002-84-2 |
| Linear Formula | C14H11 Cl2 NO3 |
| MDL Number | MFCD01672033 |
| Synonym | 2-[(2,6-Dichlorophenyl)amino]-5-hydroxybenzeneacetic acid |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 312.15 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Analytical Standard |