missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-p-naphthoquinone, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 161.0°C to 163.0°C |
| Color | Brown to Orange |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Beilstein | 08, 308 |
| Merck Index | 15, 5317 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Toxic if swallowed. May cause respiratory irritation. Causes skin irritation. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye prot |
| EINECSNumber | 207-567-5 |
| RTECSNumber | QJ5775000 |
RUO – Research Use Only