missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7131124
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15861G 1 g
H15865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15861G Supplier TCI America Supplier No. H15861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 578-22-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD09833184
InChI Key ZIOYQUNKXJQXQY-UHFFFAOYSA-N
Synonym 5-hydroxy-2-methyl-benzoic acid, 5,2-cresotic acid, 5-hydroxy-o-toluic acid, benzoic acid, 5-hydroxy-2-methyl, 2-methyl-5-hydroxybenzoic acid, acmc-1b0c9, ksc494s9h, 5-hydroxy-2-methylbenzoic acid
PubChem CID 235188
IUPAC Name 5-hydroxy-2-methylbenzoic acid
SMILES CC1=CC=C(O)C=C1C(O)=O

Specifications

Melting Point 186°C
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-2-methylbenzoic Acid
TSCA No
Recommended Storage Refrigerator