missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-adamantanone 98.0+%, TCI America™

Catalog No. H09621G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09621G 1 g
H09625G 5 g
H096225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H09621G Supplier TCI America Supplier No. H09621G

Chemical Identifiers

CAS 20098-14-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00192211
InChI Key TZBDEVBNMSLVKT-UHFFFAOYSA-N
Synonym idramantone, 5-hydroxy-2-adamantanone, kemantane, 5-hydroxy-adamantan-2-one, kemantan, 5-hydroxytricyclo 3.3.1.1 3,7 decanone, tricyclo 3.3.1.13,7 decanone, 5-hydroxy, idramantone inn, 4-oxo-1-adamantanol
PubChem CID 64184
ChEBI CHEBI:48581
IUPAC Name 5-hydroxyadamantan-2-one
SMILES C1C2CC3CC(C2)(CC1C3=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-2-adamantanone