missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-1,4-naphthoquinone 96.0+%, TCI America™

Supplier:  TCI America H02865G

 View more versions of this product

Catalog No. H02865G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Hydroxy-1,4-naphthoquinone
155°C
MFCD00001684
2811
KQPYUDDGWXQXHS-UHFFFAOYSA-N
5-hydroxy-1,4-dihydronaphthalene-1,4-dione
3806
174.16
Crystalline Powder
481-39-0
C10H6O3
5 g
juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
OC1=CC=CC2=C1C(=O)C=CC2=O
174.16
CHEBI:15794
≥96.0% (GC)

Chemical Identifiers

481-39-0
174.16
KQPYUDDGWXQXHS-UHFFFAOYSA-N
3806
5-hydroxy-1,4-dihydronaphthalene-1,4-dione
C10H6O3
MFCD00001684
juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
CHEBI:15794
OC1=CC=CC2=C1C(=O)C=CC2=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0375

RTECSNumber : QJ5775000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions