missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-1,4-naphthoquinone 96.0+%, TCI America™

Catalog No. H02861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02861G 1 g
H02865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02861G Supplier TCI America Supplier No. H02861G

Chemical Identifiers

CAS 481-39-0
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.16
MDL Number MFCD00001684
InChI Key KQPYUDDGWXQXHS-UHFFFAOYSA-N
Synonym juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
PubChem CID 3806
ChEBI CHEBI:15794
IUPAC Name 5-hydroxy-1,4-dihydronaphthalene-1,4-dione
SMILES OC1=CC=CC2=C1C(=O)C=CC2=O

Specifications

Melting Point 155°C
Quantity 1 g
UN Number 2811
Formula Weight 174.16
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-1,4-naphthoquinone
EINECSNumber (4)-0375
RTECSNumber QJ5775000
TSCA No