missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 5-Hydroxy-1,4-naphthoquinone
Description
5-Hydroxy-1,4-naphthoquinone
Specifications
Specifications
| CAS | 481-39-0 |
| Melting Point | 161°C to 163°C (lit.) |
| Molecular Formula | C10H6O3 |
| Linear Formula | C10H6O3 |
| MDL Number | MFCD00001684 |
| Quantity | 1 g |
| Synonym | Juglone |
| Molecular Weight (g/mol) | 174.15 |
| Percent Purity | 97% |